Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193609.1 | NP_001180538.1 | 4 | 633 | ||
NM_001193610.1 | NP_001180539.1 | 4 | 633 | |||
NM_001193611.1 | NP_001180540.1 | 4 | 633 | |||
NM_001193612.1 | NP_001180541.1 | 5 | 729 | |||
NM_016261.3 | NP_057345.2 | 4 | 633 | |||
XM_005257425.1 | XP_005257482.1 | 3 | 412 | |||
XM_011524871.2 | XP_011523173.1 | 4 | 631 | |||
XM_017024716.1 | XP_016880205.1 | 4 | 631 | |||
GenBank mRNA | AB030837.1 | - | 3 | 346 | ||
AB031044.1 | - | 3 | 346 | |||
AB031045.1 | - | 3 | 346 | |||
AF201333.1 | - | 3 | 345 | |||
AK022771.1 | - | 5 | 523 | |||
AK298492.1 | - | 4 | 546 | |||
AK315172.1 | - | 5 | 725 | |||
BC000258.1 | - | 4 | 608 | 3358263 | ||
BG682935.1 | - | 4 | 637 |