Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031848.1 | NP_001027018.1 | 4 | 546 | ||
NM_001276490.1 | NP_001263419.1 | 3 | 368 | |||
NM_002640.3 | NP_002631.3 | 4 | 546 | |||
NM_198833.1 | NP_942130.1 | 4 | 563 | |||
XM_006722484.2 | XP_006722547.1 | 4 | 494 | |||
XM_011526025.1 | XP_011524327.1 | 4 | 547 | |||
XM_017025790.1 | XP_016881279.1 | 4 | 547 | |||
XM_017025791.1 | XP_016881280.1 | 4 | 491 | |||
XM_017025792.1 | XP_016881281.1 | 3 | 313 | |||
GenBank mRNA | AK300391.1 | - | 3 | 316 | ||
AK313782.1 | - | 4 | 463 | |||
AL708543.1 | - | 4 | 511 | |||
BC034528.1 | - | 4 | 456 | 4780455 | ||
BQ435307.1 | - | 4 | 569 | |||
BX571754.1 | - | 4 | 454 | |||
DC384755.1 | - | 3 | 302 | |||
L40377.1 | - | 4 | 477 | |||
U92984.1 | - | 4 | 455 |