Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320509.1 | NP_001307438.1 | 12 | 1721 | ||
NM_016437.2 | NP_057521.1 | 11 | 1694 | |||
XM_011524614.2 | XP_011522916.1 | 10 | 1629 | |||
XM_017024453.1 | XP_016879942.1 | 10 | 1609 | |||
XM_017024454.1 | XP_016879943.1 | 11 | 2880 | |||
XM_017024455.1 | XP_016879944.1 | 11 | 1708 | |||
XM_017024456.1 | XP_016879945.1 | 11 | 2957 | |||
XM_017024457.1 | XP_016879946.1 | 12 | 1522 | |||
GenBank mRNA | AF225971.1 | - | 11 | 1687 | ||
AK022324.1 | - | 11 | 1637 | |||
AK225134.1 | - | 11 | 1653 | |||
BC009670.2 | - | 11 | 1546 | 3891214 | ||
BC051890.1 | - | 11 | 1637 | 5726689 | ||
BC108739.1 | - | 11 | 1655 | 5242733 | ||
BE727957.1 | - | 3 | 559 |