Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198559.1 | NP_001185488.1 | 4 | 543 | ||
NM_016544.2 | NP_057628.1 | 4 | 543 | |||
XM_011532902.1 | XP_011531204.1 | 4 | 518 | |||
XM_017004269.1 | XP_016859758.1 | 4 | 574 | |||
XM_017004270.1 | XP_016859759.1 | 4 | 434 | |||
GenBank mRNA | AF178983.1 | - | 4 | 377 | ||
AK294726.1 | - | 4 | 327 | |||
AL137731.1 | - | 5 | 566 | |||
AY094594.1 | - | 4 | 353 | |||
BC034049.1 | - | 4 | 404 | 4815297 |