Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198559.1 | NP_001185488.1 | 6 | 3480 | ||
NM_016544.2 | NP_057628.1 | 7 | 3641 | |||
XM_017004269.1 | XP_016859758.1 | 7 | 3672 | |||
XM_017004270.1 | XP_016859759.1 | 7 | 3532 | |||
GenBank mRNA | AL117490.1 | - | 1 | 2463 | ||
BC034049.1 | - | 7 | 3502 | 4815297 |