Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206528.2 | NP_001193457.1 | 6 | 1116 | ||
NM_018058.6 | NP_060528.3 | 6 | 1116 | |||
XM_005269938.4 | XP_005269995.1 | 6 | 909 | |||
XM_011539917.1 | XP_011538219.1 | 6 | 909 | |||
XM_017016366.1 | XP_016871855.1 | 6 | 909 | |||
XM_017016367.1 | XP_016871856.1 | 6 | 909 | |||
GenBank mRNA | AJ276171.1 | - | 6 | 909 | ||
AJ279016.1 | - | 5 | 736 | |||
AJ421515.1 | - | 5 | 736 | |||
AK001182.1 | - | 4 | 448 | |||
AK057190.1 | - | 6 | 1116 | |||
AK292084.1 | - | 6 | 878 | |||
BC034245.1 | - | 4 | 378 | 4178363 |