Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC8 SIRNA Assays ›
Gene Symbol:
HDAC8
Gene Name:
histone deacetylase 8
Gene Aliases:
CDA07, CDLS5, HD8, HDACL1, MRXS6, RPD3, WTS
Chromosome Location:
Chr.X: 72329516 - 72573103 on Build GRCh38
UniGene ID:
Hs.310536
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
120598
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166418.1
NP_001159890.1
2
489
NM_001166419.1
NP_001159891.1
2
489
NM_001166420.1
NP_001159892.1
2
489
NM_001166422.1
NP_001159894.1
2
489
NM_001166448.1
NP_001159920.1
2
489
NM_018486.2
NP_060956.1
2
489
NR_051952.1
2
489
XM_011530986.2
XP_011529288.2
2
484
XM_011530987.2
XP_011529289.2
2
484
XM_017029640.1
XP_016885129.1
2
484
XM_017029641.1
XP_016885130.1
2
484
XM_017029642.1
XP_016885131.1
2
482
XM_017029643.1
XP_016885132.1
2
484
XM_017029644.1
XP_016885133.1
2
484
XM_017029645.1
XP_016885134.1
2
484
XM_017029646.1
XP_016885135.1
2
164
XM_017029647.1
XP_016885136.1
2
484
GenBank mRNA
AF212246.1
-
2
205
AF230097.1
-
2
202
AF245664.1
-
2
156
AJ277724.1
-
2
189
AK074326.1
-
2
434
AK294903.1
-
2
203
AK296641.1
-
2
439
AK298767.1
-
2
229
AK300895.1
-
2
186
AY429539.1
-
2
164
BC050433.1
-
2
180
5761745
BP229332.1
-
2
465
BQ189619.1
-
2
239
BQ632790.1
-
2
201
DC352390.1
-
2
490
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
chromatin assembly or disassembly
transcription, DNA-templated
sister chromatid cohesion
chromatin modification
negative regulation of protein ubiquitination
regulation of protein stability
regulation of telomere maintenance
histone H3 deacetylation
regulation of cohesin loading
histone deacetylase activity
protein binding
transcription factor binding
Hsp70 protein binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
metal ion binding
Hsp90 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c48f4b768-29k6b:80/10.239.166.46:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0