Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166418.1 | NP_001159890.1 | 3 | 576 | ||
NM_001166419.1 | NP_001159891.1 | 5 | 849 | |||
NM_001166448.1 | NP_001159920.1 | 3 | 576 | |||
NM_018486.2 | NP_060956.1 | 5 | 849 | |||
NR_051952.1 | 4 | 707 | ||||
XM_011530986.2 | XP_011529288.2 | 5 | 844 | |||
XM_011530987.2 | XP_011529289.2 | 5 | 844 | |||
XM_017029640.1 | XP_016885129.1 | 5 | 844 | |||
XM_017029641.1 | XP_016885130.1 | 5 | 844 | |||
XM_017029642.1 | XP_016885131.1 | 3 | 569 | |||
XM_017029644.1 | XP_016885133.1 | 3 | 571 | |||
XM_017029646.1 | XP_016885135.1 | 4 | 393 | |||
XM_017029647.1 | XP_016885136.1 | 5 | 844 | |||
GenBank mRNA | AF230097.1 | - | 5 | 562 | ||
AF245664.1 | - | 5 | 516 | |||
AJ277724.1 | - | 5 | 549 | |||
AK074326.1 | - | 5 | 794 | |||
AK294903.1 | - | 3 | 290 | |||
AK296641.1 | - | 3 | 526 | |||
AK298767.1 | - | 4 | 447 | |||
AK300895.1 | - | 5 | 546 | |||
AY429539.1 | - | 4 | 393 | |||
BC050433.1 | - | 5 | 540 | 5761745 | ||
BQ632790.1 | - | 3 | 288 |