Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002646.3 | NP_002637.3 | 34 | 5870 | ||
XM_005245257.2 | XP_005245314.1 | 33 | 5935 | |||
XM_005245258.3 | XP_005245315.1 | 34 | 5854 | |||
XM_011509630.2 | XP_011507932.1 | 35 | 6729 | |||
XM_011509631.2 | XP_011507933.1 | 34 | 6046 | |||
XM_017001473.1 | XP_016856962.1 | 34 | 6865 | |||
XM_017001474.1 | XP_016856963.1 | 34 | 6068 | |||
XM_017001475.1 | XP_016856964.1 | 32 | 4816 | |||
GenBank mRNA | AB210006.1 | - | 32 | 5424 | ||
BC132876.1 | - | 34 | 5842 | 40146898 | ||
BC136645.1 | - | 34 | 5842 | 9020635 | ||
BC144342.1 | - | 35 | 5758 | 9052862 | ||
CR749201.1 | - | 4 | 648 | |||
Y11312.1 | - | 34 | 5906 |