Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021205.5 | NP_067028.1 | 3 | 1156 | ||
NR_037962.1 | 3 | 614 | ||||
GenBank mRNA | AB051826.1 | - | 3 | 569 | ||
AB074878.1 | - | 3 | 629 | |||
AB209511.1 | - | 4 | 688 | |||
AF211836.1 | - | 3 | 1112 | |||
AF251701.1 | - | 3 | 349 | |||
AF282258.1 | - | 3 | 729 | |||
AF378087.1 | - | 3 | 490 | |||
AK289971.1 | - | 3 | 742 | |||
BC040076.1 | - | 3 | 737 | 5768957 |