Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021205.5 | NP_067028.1 | 3 | 1800 | ||
NR_037962.1 | 3 | 1258 | ||||
GenBank mRNA | AB209511.1 | - | 4 | 1332 | ||
AF211836.1 | - | 3 | 1756 | |||
AF282258.1 | - | 3 | 1372 | |||
AK289971.1 | - | 3 | 1386 | |||
BC040076.1 | - | 3 | 1381 | 5768957 |