Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021205.5 | NP_067028.1 | 3 | 3076 | ||
NR_037962.1 | 3 | 2534 | ||||
GenBank mRNA | AB209511.1 | - | 4 | 2608 | ||
AF211836.1 | - | 3 | 3034 | |||
AK001478.1 | - | 1 | 1132 | |||
BC040076.1 | - | 3 | 2657 | 5768957 |