Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022066.3 | NP_071349.3 | 18 | 4386 | ||
XM_005257567.1 | XP_005257624.1 | 18 | 4373 | |||
XM_017024934.1 | XP_016880423.1 | 18 | 4298 | |||
XM_017024935.1 | XP_016880424.1 | 18 | 4295 | |||
GenBank mRNA | AB051521.1 | - | 18 | 4386 | ||
AK000926.1 | - | 1 | 621 | |||
AK022940.1 | - | 5 | 1884 | |||
AK024657.1 | - | 3 | 1385 | |||
AK025999.1 | - | 10 | 2865 | |||
AL832432.1 | - | 11 | 3311 | |||
BC004525.2 | - | 3 | 1201 | |||
BC022237.1 | - | 5 | 1904 | |||
BC025977.1 | - | 5 | 1785 | |||
BC036820.1 | - | 13 | 3522 | |||
BC051868.2 | - | 10 | 2729 | 6162673 |