Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171864.1 | NP_001165335.1 | 4 | 496 | ||
NM_024803.2 | NP_079079.1 | 4 | 616 | |||
GenBank mRNA | AK025318.1 | - | 4 | 618 | ||
AK296616.1 | - | 4 | 494 | |||
BC098247.1 | - | 4 | 489 | 40005930 | ||
BC099716.1 | - | 4 | 609 | 40005934 | ||
BC105634.1 | - | 4 | 609 | 40005931 |