Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030773.3 | NP_110400.1 | 3 | 501 | ||
XM_017028085.1 | XP_016883574.1 | 3 | 695 | |||
GenBank mRNA | AJ292757.1 | - | 3 | 501 | ||
BC033679.1 | - | 3 | 334 | 5180793 | ||
BI821931.1 | - | 3 | 334 |