Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030773.3 | NP_110400.1 | 4 | 2746 | ||
XM_017028085.1 | XP_016883574.1 | 4 | 2940 | |||
GenBank mRNA | AJ292757.1 | - | 4 | 2745 | ||
BC033679.1 | - | 4 | 2577 | 5180793 |