Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031296.2 | NP_112586.1 | 2 | 690 | ||
XM_011532299.1 | XP_011530601.1 | 3 | 786 | |||
GenBank mRNA | AF350420.1 | - | 2 | 412 | ||
AK313685.1 | - | 2 | 350 | |||
AL136904.1 | - | 2 | 690 | |||
BC036064.1 | - | 2 | 322 | 5288066 | ||
BC111977.1 | - | 2 | 342 | 8327445 |