Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257137.2 | NP_001244066.1 | 10 | 1122 | ||
NM_001257138.2 | NP_001244067.1 | 8 | 857 | |||
NM_001324197.1 | NP_001311126.1 | 10 | 1143 | |||
NM_001324198.1 | NP_001311127.1 | 9 | 1020 | |||
NM_031483.6 | NP_113671.3 | 9 | 999 | |||
XM_005260570.1 | XP_005260627.1 | 8 | 915 | |||
XM_011529079.1 | XP_011527381.1 | 8 | 935 | |||
XM_017028089.1 | XP_016883578.1 | 9 | 955 | |||
XM_017028090.1 | XP_016883579.1 | 10 | 1075 | |||
XM_017028091.1 | XP_016883580.1 | 8 | 870 | |||
XM_017028092.1 | XP_016883581.1 | 10 | 1075 | |||
GenBank mRNA | AB056663.2 | - | 9 | 909 | ||
AB209747.1 | - | 1 | 16 | |||
AF038564.1 | - | 4 | 417 | |||
AF095745.1 | - | 7 | 792 | |||
AK297809.1 | - | 9 | 817 | |||
AK304090.1 | - | 8 | 816 | |||
AK315212.1 | - | 9 | 958 | |||
BC011571.1 | - | 10 | 1045 | 4560366 |