Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159286.1 | NP_001152758.1 | 13 | 1707 | ||
NM_032019.5 | NP_114408.3 | 14 | 1767 | |||
GenBank mRNA | AF393962.1 | - | 14 | 1394 | ||
AF407272.1 | - | 14 | 1394 | |||
AF407273.1 | - | 13 | 1334 | |||
AF426160.1 | - | 14 | 1747 | |||
AL512711.1 | - | 2 | 657 | |||
BC032049.1 | - | 14 | 1580 | |||
BC094734.1 | - | 12 | 1435 | |||
BC125083.1 | - | 14 | 1441 | 40117772 | ||
CR456465.1 | - | 14 | 1450 |