Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032515.4 | NP_115904.1 | 5 | 2005 | ||
XM_011511696.2 | XP_011509998.1 | 5 | 1809 | |||
XM_017004775.1 | XP_016860264.1 | 6 | 2107 | |||
XM_017004776.1 | XP_016860265.1 | 6 | 2033 | |||
GenBank mRNA | BC006203.2 | - | 7 | 1691 | ||
BG118500.1 | - | 1 | 352 | |||
BU855574.1 | - | 1 | 837 | |||
CN279174.1 | - | 1 | 486 |