Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032918.2 | NP_116307.1 | 3 | 493 | ||
XM_017020121.1 | XP_016875610.1 | 4 | 999 | |||
GenBank mRNA | AF339750.1 | - | 2 | 236 | ||
AK313881.1 | - | 3 | 433 | |||
BC007997.1 | - | 3 | 386 | 3357341 |