Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004985.4 | NP_004976.2 | 5 | 2773 | ||
NM_033360.3 | NP_203524.1 | 6 | 2897 | |||
XM_011520653.2 | XP_011518955.1 | 5 | 2765 | |||
XM_017019293.1 | XP_016874782.1 | 6 | 3385 | |||
GenBank mRNA | BC010502.1 | - | 1 | 1215 | ||
M54968.1 | - | 5 | 2776 |