Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145311.1 | NP_001138783.1 | 9 | 2109 | ||
NM_002666.4 | NP_002657.3 | 9 | 2180 | |||
XM_005254934.4 | XP_005254991.1 | 9 | 2038 | |||
GenBank mRNA | AB005293.1 | - | 9 | 2123 | ||
BC031084.1 | - | 9 | 2123 | 5284402 |