Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031677.3 | NP_001026847.1 | 2 | 626 | ||
NM_130781.3 | NP_570137.2 | 3 | 540 | |||
NR_109789.1 | 3 | 540 | ||||
NR_109790.1 | 2 | 626 | ||||
GenBank mRNA | AF258583.1 | - | 2 | 370 | ||
AK021761.1 | - | 1 | 657 | |||
AK308155.1 | - | 3 | 495 | |||
AL833898.1 | - | 2 | 168 | |||
BC010006.2 | - | 3 | 463 | 4300377 | ||
BC015534.1 | - | 3 | 502 | 3916832 | ||
BC021263.2 | - | 2 | 586 | 4329216 | ||
BT007268.1 | - | 2 | 170 | |||
DA898352.1 | - | 3 | 537 |