Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHEBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303126.1 | NP_001290055.1 | 7 | 700 | ||
NM_144593.2 | NP_653194.1 | 7 | 632 | |||
NR_130123.1 | 6 | 564 | ||||
XM_017018794.1 | XP_016874283.1 | 7 | 1289 | |||
XM_017018795.1 | XP_016874284.1 | 7 | 1357 | |||
GenBank mRNA | AK098663.1 | - | 7 | 631 | ||
AY327412.1 | - | 7 | 632 | |||
AY509932.1 | - | 7 | 698 | |||
BC014155.2 | - | 6 | 486 | |||
BC027482.1 | - | 7 | 599 | 4396827 |