Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004841.3 | NP_004832.1 | 6 | 971 | ||
NM_170692.2 | NP_733793.2 | 8 | 2054 | |||
XM_005245622.3 | XP_005245679.1 | 6 | 1627 | |||
XM_011510166.2 | XP_011508468.1 | 8 | 2054 | |||
XM_011510167.2 | XP_011508469.1 | 8 | 2054 | |||
XM_017002849.1 | XP_016858338.1 | 8 | 4660 | |||
XM_017002850.1 | XP_016858339.1 | 8 | 4659 | |||
XM_017002851.1 | XP_016858340.1 | 8 | 4659 | |||
XM_017002852.1 | XP_016858341.1 | 8 | 4658 | |||
XM_017002853.1 | XP_016858342.1 | 8 | 2098 | |||
XM_017002854.1 | XP_016858343.1 | 6 | 1630 | |||
XM_017002855.1 | XP_016858344.1 | 6 | 1629 | |||
GenBank mRNA | AF047711.1 | - | 6 | 971 | ||
AK290308.1 | - | 6 | 914 | |||
BC110611.2 | - | 8 | 1286 | 40028914 |