Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286075.1 | NP_001273004.1 | 3 | 257 | ||
NM_004320.4 | NP_004311.1 | 5 | 608 | |||
NM_173201.3 | NP_775293.1 | 5 | 608 | |||
GenBank mRNA | AK128456.1 | - | 3 | 247 | ||
AK291314.1 | - | 5 | 537 | |||
AL706356.1 | - | 3 | 257 | |||
BC037354.1 | - | 5 | 536 | |||
BX537389.1 | - | 5 | 604 | |||
BX537784.1 | - | 5 | 606 | |||
BX647365.1 | - | 5 | 604 | |||
BX647366.1 | - | 5 | 600 | |||
BX647367.1 | - | 5 | 608 | |||
BX648544.1 | - | 5 | 605 | |||
BX649072.1 | - | 5 | 597 |