Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 4 | 2305 | ||
NM_001293213.1 | NP_001280142.1 | 5 | 1615 | |||
NM_001293214.1 | NP_001280143.1 | 3 | 2089 | |||
NM_001293215.1 | NP_001280144.1 | 4 | 2208 | |||
NM_001293216.1 | NP_001280145.1 | 4 | 2107 | |||
NM_178014.3 | NP_821133.1 | 4 | 2221 | |||
NR_120608.1 | 4 | 1622 | ||||
GenBank mRNA | AB062393.1 | - | 4 | 2049 | ||
AK098772.1 | - | 4 | 2066 | |||
BC001002.2 | - | 1 | 850 | |||
BC007605.1 | - | 4 | 2051 | 3350604 | ||
BC013374.2 | - | 4 | 2042 | 3946253 | ||
BC021909.1 | - | 4 | 2041 | 3509417 | ||
BC062532.1 | - | 4 | 1384 |