Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178822.4 | NP_849144.2 | 4 | 2782 | ||
XM_011512708.2 | XP_011511010.1 | 10 | 3591 | |||
XM_011512709.2 | XP_011511011.1 | 9 | 3322 | |||
GenBank mRNA | AY273815.1 | - | 4 | 2782 |