Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284338.1 | NP_001271267.1 | 20, 21 | 3827 | ||
NM_001284339.1 | NP_001271268.1 | 20, 21 | 3779 | |||
NM_001284340.1 | NP_001271269.1 | 19, 20 | 3776 | |||
NM_006154.3 | NP_006145.2 | 24, 25 | 2461 | |||
NM_198400.3 | NP_940682.2 | 17, 18 | 3611 | |||
XM_011521624.2 | XP_011519926.1 | 23, 24 | 2226 | |||
XM_011521625.2 | XP_011519927.1 | 25, 26 | 2399 | |||
XM_011521626.1 | XP_011519928.1 | 23, 24 | 2243 | |||
XM_011521627.1 | XP_011519929.1 | 21, 22 | 2066 | |||
GenBank mRNA | AK304394.1 | - | 24, 25 | 2327 | ||
AK307541.1 | - | 20, 21 | 2078 | |||
AL832063.1 | - | 17, 18 | 3612 | |||
AL833300.1 | - | 20, 21 | 2809 | |||
AY550969.1 | - | 24, 25 | 2340 | |||
BC035707.1 | - | 24, 25 | 2305 | |||
BC136605.1 | - | 20, 21 | 3804 | 9020595 | ||
BC144284.1 | - | 20, 21 | 3804 | 9052804 | ||
BC144285.1 | - | 20, 21 | 3756 | 9052805 | ||
BC152452.1 | - | 24, 25 | 2352 | 8862584 | ||
BC152562.1 | - | 24, 25 | 2352 | 8860509 | ||
D42055.1 | - | 24, 25 | 2352 |