Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004322.3 | NP_004313.1 | 3 | 922 | ||
NM_032989.2 | NP_116784.1 | 4 | 668 | |||
GenBank mRNA | AF021792.1 | - | 3 | 586 | ||
AK291863.1 | - | 4 | 668 | |||
BC001901.2 | - | 3 | 594 | 3537914 | ||
BC095431.1 | - | 4 | 633 | 30377088 | ||
BG748336.1 | - | 4 | 656 | |||
U66879.1 | - | 3 | 652 |