Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252039.1 | NP_001238968.1 | 7 | 1273 | ||
NM_004583.3 | NP_004574.2 | 6 | 1057 | |||
NM_201434.2 | NP_958842.1 | 7 | 1174 | |||
XM_011525090.1 | XP_011523392.1 | 6 | 996 | |||
XM_011525091.1 | XP_011523393.1 | 6 | 1120 | |||
GenBank mRNA | BC106039.1 | - | 6 | 1016 | 4938169 | |
BC114439.1 | - | 6 | 1012 | 40083572 | ||
U11293.1 | - | 5 | 892 | |||
U18420.1 | - | 6 | 952 |