Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006270.4 | NP_006261.1 | 3, 4 | 397 | ||
GenBank mRNA | AF493920.1 | - | 3, 4 | 352 | ||
BC016286.1 | - | 3, 4 | 376 | 3910343 | ||
BC016318.1 | - | 3, 4 | 401 | 4073829 | ||
BT006805.1 | - | 3, 4 | 352 | |||
CR541944.1 | - | 3, 4 | 352 | |||
CR541967.1 | - | 3, 4 | 352 |