Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002689.3 | NP_002680.2 | 8 | 1325 | ||
XM_011544877.2 | XP_011543179.1 | 8 | 1264 | |||
XM_011544878.2 | XP_011543180.1 | 8 | 1264 | |||
XM_011544879.1 | XP_011543181.1 | 8 | 1264 | |||
XM_011544881.2 | XP_011543183.1 | 8 | 1264 | |||
XM_017017452.1 | XP_016872941.1 | 8 | 1264 | |||
GenBank mRNA | AK025315.1 | - | 8 | 1188 | ||
AK094569.1 | - | 1 | 664 | |||
AK297842.1 | - | 7 | 1090 | |||
AK307831.1 | - | 8 | 1215 | |||
BC001347.2 | - | 8 | 1226 | 2822514 | ||
BC002990.1 | - | 8 | 1226 | 2822514 | ||
BC018813.2 | - | 8 | 1226 | |||
BC018814.2 | - | 8 | 1226 | |||
CR456737.1 | - | 8 | 884 | |||
L24559.1 | - | 8 | 948 |