Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002690.2 | NP_002681.1 | 14 | 1257 | ||
XM_005273535.3 | XP_005273592.1 | 15 | 1359 | |||
XM_005273536.3 | XP_005273593.1 | 14 | 1272 | |||
XM_005273537.3 | XP_005273594.1 | 13 | 1167 | |||
XM_005273538.2 | XP_005273595.1 | 14 | 1269 | |||
XM_005273539.2 | XP_005273596.1 | 13 | 1164 | |||
XM_005273540.4 | XP_005273597.1 | 10 | 1018 | |||
XM_006716353.2 | XP_006716416.1 | 11 | 903 | |||
XM_017013583.1 | XP_016869072.1 | 12 | 1161 | |||
XM_017013584.1 | XP_016869073.1 | 11 | 1054 | |||
GenBank mRNA | AK223537.1 | - | 14 | 1256 | ||
AK294025.1 | - | 10 | 976 | |||
BC100288.1 | - | 14 | 1221 | 6151069 | ||
BC106909.1 | - | 14 | 1183 | 40031807 | ||
D29013.1 | - | 14 | 1200 | |||
M13140.1 | - | 14 | 1200 |