Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001975.1 | NP_001001975.1 | 1 | 217 | ||
NM_001687.4 | NP_001678.1 | 1 | 217 | |||
GenBank mRNA | AK094357.1 | - | 1 | 199 | ||
BC002389.2 | - | 1 | 217 | 2819749 | ||
BC004426.1 | - | 1 | 217 | 2819749 | ||
BC018079.1 | - | 1 | 119 | 4798666 | ||
BC018817.2 | - | 1 | 217 | |||
CR542218.1 | - | 1 | 116 | |||
S87916.1 | - | 1 | 199 | |||
S87918.1 | - | 1 | 199 | |||
X63422.1 | - | 1 | 199 | |||
X63423.1 | - | 1 | 199 |