Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003806.2 | NP_003797.1 | 2 | 491 | ||
NR_073189.1 | 5 | 831 | ||||
XM_017020146.1 | XP_016875635.1 | 5 | 799 | |||
GenBank mRNA | BE674890.1 | - | 5 | 492 | ||
D83699.1 | - | 2 | 96 | |||
U76376.1 | - | 2 | 491 |