Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019034.2 | NP_061907.2 | 3 | 339 | ||
GenBank mRNA | AF239923.1 | - | 3 | 313 | ||
AK000254.1 | - | 3 | 305 | |||
AK289482.1 | - | 3 | 913 | |||
BC018208.1 | - | 3 | 318 | 3864426 | ||
BG574506.1 | - | 4 | 621 | |||
BQ053707.1 | - | 3 | 320 | |||
BU527972.1 | - | 3 | 321 |