Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 2, 3 | 558 | ||
NM_001293213.1 | NP_001280142.1 | 2, 3 | 474 | |||
NM_001293215.1 | NP_001280144.1 | 2, 3 | 461 | |||
NM_001293216.1 | NP_001280145.1 | 2, 3 | 360 | |||
NM_178014.3 | NP_821133.1 | 2, 3 | 474 | |||
NR_120608.1 | 2, 3 | 474 | ||||
GenBank mRNA | AB062393.1 | - | 2, 3 | 300 | ||
AF070561.1 | - | 2, 3 | 295 | |||
AF070593.1 | - | 2, 3 | 295 | |||
AF070600.1 | - | 2, 3 | 312 | |||
AF141349.1 | - | 2, 3 | 312 | |||
AK098772.1 | - | 2, 3 | 319 | |||
AK297489.1 | - | 2, 3 | 228 | |||
AK297638.1 | - | 2, 3 | 323 | |||
AK298889.1 | - | 2, 3 | 364 | |||
AK302314.1 | - | 2, 3 | 427 | |||
AK316265.1 | - | 2, 3 | 231 | |||
AK316316.1 | - | 2, 3 | 356 | |||
BC001938.1 | - | 2, 3 | 299 | 2820585 | ||
BC002347.2 | - | 2, 3 | 304 | 2964461 | ||
BC005838.2 | - | 2, 3 | 299 | 2820585 | ||
BC007605.1 | - | 2, 3 | 304 | 3350604 | ||
BC013374.2 | - | 2, 3 | 295 | 3946253 | ||
BC019924.1 | - | 2, 3 | 292 | 4998571 | ||
BC020946.1 | - | 2, 3 | 333 | 4795692 | ||
BC021909.1 | - | 2, 3 | 294 | 3509417 | ||
BC062532.1 | - | 2, 3 | 292 | |||
BC070326.1 | - | 2, 3 | 310 | 6698679 | ||
BC103746.1 | - | 2, 3 | 317 | 6503977 | ||
BE245819.1 | - | 2, 3 | 328 | |||
BU557299.1 | - | 2, 3 | 474 | |||
DC353572.1 | - | 2, 3 | 462 | |||
DC405540.1 | - | 2, 3 | 271 | |||
V00599.1 | - | 1, 2 | 147 |