Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015276.1 | NP_056091.1 | 13 | 2521 | ||
XM_005256575.2 | XP_005256632.1 | 13 | 2235 | |||
GenBank mRNA | AB028986.1 | - | 13 | 2522 | ||
AK025331.1 | - | 1 | 43 | |||
BC007196.1 | - | 6 | 1263 | |||
BC013364.2 | - | 1 | 80 | |||
BX640815.1 | - | 12 | 2104 |