Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006001.2 | NP_005992.1 | 3 | 358 | ||
GenBank mRNA | BC011721.2 | - | 3 | 301 | 4025942 | |
DB026664.1 | - | 3 | 358 |