Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002705.4 | NP_002696.3 | 22 | 3889 | ||
XM_006720902.3 | XP_006720965.1 | 22 | 3853 | |||
XM_017023374.1 | XP_016878863.1 | 22 | 3901 | |||
XM_017023375.1 | XP_016878864.1 | 22 | 3913 | |||
GenBank mRNA | AB011140.1 | - | 13 | 2811 | ||
AF001691.1 | - | 22 | 3889 | |||
AF013717.1 | - | 22 | 3810 | |||
AK295623.1 | - | 10 | 2330 | |||
BC114620.1 | - | 22 | 3849 | 40070172 |