Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4RL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178570.2 | NP_848665.1 | 2 | 612 | ||
GenBank mRNA | AF532858.1 | - | 2 | 283 | ||
AX751514.1 | - | 2 | 283 | |||
AY250221.1 | - | 2 | 283 | |||
BC113673.1 | - | 2 | 305 | 8322725 | ||
BC117276.1 | - | 2 | 305 | 40125827 | ||
BC143818.1 | - | 2 | 305 | 9052333 | ||
DQ864979.1 | - | 2 | 283 |