Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002556.2 | NP_002547.1 | 14 | 3160 | ||
GenBank mRNA | AF185696.1 | - | 14 | 3160 | ||
BC011581.1 | - | 14 | 2748 | 4560111 | ||
BC063121.1 | - | 8 | 1519 |