Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004104.4 | NP_004095.4 | 3 | 293 | ||
XM_011523538.2 | XP_011521840.1 | 3 | 361 | |||
GenBank mRNA | AB209988.1 | - | 3 | 288 | ||
AY451392.1 | - | 2 | 176 | |||
BC063242.1 | - | 3 | 293 | 6172538 | ||
U26644.1 | - | 2 | 176 | |||
U29344.1 | - | 3 | 299 |