Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004378.2 | NP_004369.1 | 3, 4 | 461 | ||
GenBank mRNA | AB593091.1 | - | 3, 4 | 435 | ||
AL110113.1 | - | 3, 4 | 345 | |||
AU132726.1 | - | 3, 4 | 461 | |||
BC022069.1 | - | 3, 4 | 437 | 4797853 | ||
CR457017.1 | - | 3, 4 | 356 | |||
S74445.1 | - | 3, 4 | 430 |