Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005564.4 | NP_005555.2 | 4 | 475 | ||
GenBank mRNA | AK301694.1 | - | 4 | 452 | ||
AK316217.1 | - | 4 | 452 | |||
BC033089.1 | - | 4 | 439 | 5421124 | ||
BP272828.1 | - | 4 | 452 | |||
BX644845.1 | - | 4 | 488 | |||
CA454137.1 | - | 4 | 437 | |||
CR542092.1 | - | 4 | 379 | |||
EU644752.1 | - | 4 | 385 | |||
S75256.1 | - | 4 | 319 | |||
X83006.1 | - | 4 | 442 |