Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174060.1 | NP_001167531.1 | 3 | 1181 | ||
NM_017784.4 | NP_060254.2 | 3 | 1181 | |||
XM_005264843.3 | XP_005264900.1 | 3 | 957 | |||
XM_005264844.4 | XP_005264901.1 | 3 | 957 | |||
XM_011533326.2 | XP_011531628.1 | 3 | 334 | |||
XM_017005667.1 | XP_016861156.1 | 3 | 847 | |||
XM_017005668.1 | XP_016861157.1 | 3 | 824 | |||
XM_017005669.1 | XP_016861158.1 | 3 | 353 | |||
GenBank mRNA | AF346291.1 | - | 3 | 627 | ||
AF392451.1 | - | 4 | 891 | |||
AK000370.1 | - | 1 | 57 | |||
AK304067.1 | - | 3 | 634 | |||
AL833247.1 | - | 3 | 284 | |||
BC003168.1 | - | 3 | 585 | 2905981 |