Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166660.1 | NP_001160132.1 | 6 | 2505 | ||
NM_001321276.1 | NP_001308205.1 | 7 | 2052 | |||
NM_018977.3 | NP_061850.2 | 7 | 2565 | |||
NM_181303.1 | NP_851820.1 | 8 | 2625 | |||
XM_006724662.3 | XP_006724725.2 | 9 | 2440 | |||
XM_006724663.3 | XP_006724726.1 | 9 | 2632 | |||
XM_011530974.2 | XP_011529276.1 | 8 | 2554 | |||
GenBank mRNA | AB040913.1 | - | 5 | 1795 | ||
AF217411.1 | - | 7 | 2526 | |||
AK074814.1 | - | 2 | 864 | |||
AK296217.1 | - | 5 | 1860 | |||
AK299669.1 | - | 7 | 2036 | |||
AK314699.1 | - | 6 | 2342 | |||
BC051715.1 | - | 7 | 2495 | 5258980 | ||
BX641059.1 | - | 7 | 2048 | |||
GQ489207.1 | - | 7 | 2291 | |||
KJ534889.1 | - | 6 | 2231 | |||
KJ534890.1 | - | 5 | 2171 | |||
KJ534891.1 | - | 7 | 2291 | |||
KJ534977.1 | - | 5 | 2171 | |||
KJ535034.1 | - | 5 | 1441 | |||
KJ535040.1 | - | 6 | 2089 | |||
KU178651.1 | - | 6 | 2231 |