Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128304.1 | NP_001121776.1 | 11 | 1379 | ||
NM_001128305.1 | NP_001121777.1 | 9 | 1341 | |||
NM_001128306.1 | NP_001121778.1 | 7 | 983 | |||
NM_001177304.1 | NP_001170775.1 | 6 | 955 | |||
NM_020353.2 | NP_065086.2 | 9 | 1253 | |||
XM_005247654.2 | XP_005247711.1 | 10 | 1280 | |||
XM_005247655.2 | XP_005247712.1 | 10 | 1197 | |||
XM_011513031.2 | XP_011511333.1 | 9 | 1139 | |||
XM_017006921.1 | XP_016862410.1 | 8 | 1010 | |||
GenBank mRNA | AF199023.1 | - | 9 | 1157 | ||
AK128442.1 | - | 6 | 855 | |||
AK290214.1 | - | 9 | 1281 | |||
AK292815.1 | - | 11 | 1279 | |||
AL833760.1 | - | 9 | 1167 | |||
BC028354.2 | - | 9 | 1253 | 4831340 |